Order Entry
Germany
ContactUsLinkComponent
AEROSIL® R 812S
AEROSIL® R 812S
Catalog # OVERAEROSILR812S
CAS Number:  
AEROSIL® R 812S
Catalog # OVERAEROSILR812S
CAS Number:  

Specifications

  • Pk:
    10 kg
  • Seq:
    4158

Specifications

About this item

 :
CAS Number:112945-52-5
 :7631-86-9
MDL Number:MFCD00011232