Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromatography Sorbents
HPLC Sorbents
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packing
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packing
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packing
Catalog # 7289-05
Supplier:
Avantor
CAS Number:
Expand image
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packing
Catalog # 7289-05
Supplier:
Avantor
CAS Number:
Specifications
Particle Size:
15 µm
Pk:
500 g
Description:
Wide-pore prep packing
Specifications
About this item
Product Details
Product Details