Order Entry
Germany
ContactUsLinkComponent
 
Ion exchange chromatography columns, Capto™ HiRes
 :  Cytiva
 
undefined
Ion exchange chromatography columns, Capto™ HiRes
 :  Cytiva

 

  • Catalog No:
  • 29-2758-79
  • 29-2758-78
  • 29-2758-77
  • 29-2758-81
  • Max. pressure:
  • 40 bar
  • 40 bar
  • 40 bar
  • 40 bar
  • pH range:
  • 2 - 12
  • 2 - 12
  • 2 - 12
  • 2 - 12
  • Storage:
  • 4 to 30 °C
  • 4 to 30 °C
  • 4...30 °C
  • 4 to 30 °C