J.T.Baker® BAKERBOND®, Wide-Pore Carboxy-Sulfon Ion Exchnge Media
Supplier: Avantor
Certificates
About this item
Prep-scale packing is intended for use at low pressure with a peristaltic pump.
- Weak-strong cation exchanger
Prep-scale packing is intended for use at low pressure with a peristaltic pump.