Order Entry
Germany
ContactUsLinkComponent
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packing
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packing
  7248-00
 :  Avantor
 :  
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packing
  7248-00
 :  Avantor
 :  

 

  • Particle Size:
    40 µm
  • Pk:
    100 g
  • Pore size:
    275 Å
  • Description:
    Octadecyl (C18), BAKERBOND®
  • Seq:
    01

 

 

Prep-scale packing is intended for use at low pressure with a peristaltic pump.