- SNR:35 db
- Colour:Green/black
- Description:Earmuffs, Peltor™ Optime III™, with headband
- Pk:1
The earmuffs are designed specifically for heavy noise and combine effective protection with the ultimate in comfort. The model H540AV with headband is in Hi-VIZ design for high visibility. The advantage like conversations and warning signals are still easy to understand. An acoustic connection between the capsule interior and the space between the shells also achieves optimal low-frequency insulation.
- Double capsule construction for highest insulation performance
- Filled with a unique combination of liquid and foam
- Perfect adaptation to the head shape
- Very good protection at high volumes
- Very high security
- Compatibility with other PSA
The generous space under the capsules minimises heat build-up and perspiration and provides a comfortable fit. For a consistently low contact pressure even with permanent use, the low bracket and the two-point hanger made of stainless steel.
Matching hygiene sets ensure a hygienic and economical use of the earmuffs. For a consistently low contact pressure even with permanent use, the low bracket and the two-point hanger made of stainless steel. The bracket is variably adjustable and adapts perfectly to any head shape thanks to the ergonomic shape. It uses high noise levels wherever high visibility is required, such as roadworks and airports. Matching hygiene sets ensure a hygienic and economical use of the earmuffs.
SNR: 35 dB