Nuclear Fast Red, Gurr™ for microscopical staining
Wechseln zu:
Product Details & Documents
Synonyme: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
- CAS-Nummer:6409-77-4
- Formel::C₁₄H₈NNaO₇S
- MDL-Nummer:MFCD00078493
- Molekulargewicht:357.28 g/mol
Documents
Spezifikationen:
Spezifikation Testergebnisse
- Dye content (spectrophotometrically):Min. 90 %
- Identity (Sodium):Passes test
- Identity (UV/VIS-Spectrum):Passes test
- Absorption max. λ 1 (Dimethyl sulfoxide):557 --> 559 nm
- Absorption max. λ 2 (Dimethyl sulfoxide):520 --> 522 nm
- Spec.Absorptivity (λ1max; 0.02 g/l;DMSO):380 --> 422
- Spec.Absorptivity (λ2max; 0.02 g/l;DMSO):383 --> 426
- TLC-Test:Passes test
- Loss on drying (110°C):Max. 5 %
- Suitability for microscopy:Passes test



