Order Entry
Germany
ContactUsLinkComponent
 
Pesticide reference standard, Fenthion-oxon, neat, Honeywell Fluka™
Catalog # HONCR1529-10MG
CAS Number:  
Pesticide reference standard, Fenthion-oxon, neat, Honeywell Fluka™
Catalog # HONCR1529-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    6552-12-1
  • Description:
    Fenthion-oxon
  • Formula:
    C₁₀H₁₅O₄PS
  • Storage temperature:
    Ambient
  • Description:
    Fenthion-oxon

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results