Auftragserfassung
Germany
ContactUsLinkComponent
Pesticide reference standard, (S,S)-Metalaxyl Metabolite CGA 226048, neat, Honeywell Fluka™
Pesticide reference standard, (S,S)-Metalaxyl Metabolite CGA 226048, neat, Honeywell Fluka™
Catalog # HONCR1522-10MG
CAS Number:  
Pesticide reference standard, (S,S)-Metalaxyl Metabolite CGA 226048, neat, Honeywell Fluka™
Catalog # HONCR1522-10MG
CAS Number:  

Specifications

  • Packung:
    Glass bottle
  • VE:
    10 mg
  • Bezeichnung:
    (S,S)-Metalaxyl Metabolite CGA 226048
  • Formel:
    C₁₈H₂₅NO₆
  • Lagertemperatur:
    Ambient
  • Bezeichnung:
    (S,S)-Metalaxyl Metabolite CGA 226048
  • Gefrierpunkt:
    92...95 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results