Pesticide reference standard, Ethoxyquin hydrochloride, neat, Honeywell Fluka™
Catalog # HONCR1505-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:3659-01-6
- Description:Ethoxyquin hydrochloride
- Formula:C₁₄H₁₉NO · HCl
- Storage temperature:Ambient
- Description:Ethoxyquin hydrochloride
- Freezing point:175...180 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results