Order Entry
Germany
ContactUsLinkComponent
Organic analytical standards, Dichloro-isoeverninic acid, neat, reference material, Honeywell Fluka™
Organic analytical standards, Dichloro-isoeverninic acid, neat, reference material, Honeywell Fluka™
Catalog # HONCR1479-10MG
CAS Number:  
Organic analytical standards, Dichloro-isoeverninic acid, neat, reference material, Honeywell Fluka™
Catalog # HONCR1479-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    4101-80-8
  • Description:
    Dichloro-isoeverninic acid
  • Formula:
    C₉H₈Cl₂O₄
  • Storage temperature:
    Ambient
  • Description:
    Dichloro-isoeverninic acid
  • Freezing point:
    126...128 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results