Order Entry
Germany
ContactUsLinkComponent
Organic analytical standards, Zearalenone-14-O-beta-glucoside, neat, reference material, Honeywell Fluka™
Organic analytical standards, Zearalenone-14-O-beta-glucoside, neat, reference material, Honeywell Fluka™
Catalog # HONCR1442-10MG
CAS Number:  
Organic analytical standards, Zearalenone-14-O-beta-glucoside, neat, reference material, Honeywell Fluka™
Catalog # HONCR1442-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    105088-14-0
  • Description:
    Zearalenone-14-O-beta-glucoside
  • Formula:
    C₂₄H₃₂O₁₀
  • Storage temperature:
    –20 °C
  • Description:
    Zearalenone-14-O-beta-glucoside
  • Freezing point:
    211...215 °C

Specifications

About this item

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results