Order Entry
Germany
ContactUsLinkComponent
Organic analytical standards, alpha-Zearalanol-d5, neat, reference material, Honeywell Fluka™
Organic analytical standards, alpha-Zearalanol-d5, neat, reference material, Honeywell Fluka™
Catalog # HONCR1439-10MG
CAS Number:  
Organic analytical standards, alpha-Zearalanol-d5, neat, reference material, Honeywell Fluka™
Catalog # HONCR1439-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    alpha-Zearalanol-d₅
  • Melting point:
    164...174 °C
  • Formula:
    C₁₈H₂₁D₅O₅
  • Storage temperature:
    –20 °C
  • Description:
    alpha-Zearalanol-d₅
  • Boiling point:
    576 °C
  • Freezing point:
    169 - 172 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results