Pharmaceutical analytical standard, Cefacetrile-13C3, reference material, Honeywell Fluka™
Catalog # HONCR1435-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Description:Cefacetrile-¹³C₃
- Formula:C₁₀¹³C₃H₁₃N₃O₆S
- Storage temperature:Ambient
- Description:Cefacetrile-¹³C₃
- Freezing point:166...169 °C
Specifications
About this item
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results