Order Entry
Germany
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Clenbuterol-d9, reference material, Honeywell Fluka™
Catalog # HONCR1407-10MG
CAS Number:  
Pharmaceutical analytical standard, Clenbuterol-d9, reference material, Honeywell Fluka™
Catalog # HONCR1407-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    129138-58-5
  • Description:
    Clenbuterol-d₉
  • Formula:
    C₁₂H₉D₉Cl₂N₂O
  • Storage temperature:
    Ambient
  • Description:
    Clenbuterol-d₉
  • Freezing point:
    122...123 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results