Order Entry
Germany
ContactUsLinkComponent
Pesticide reference standard, 2-(Diethylamino)-6-methyl-4-pyrimidinol, neat, Honeywell Fluka™
Pesticide reference standard, 2-(Diethylamino)-6-methyl-4-pyrimidinol, neat, Honeywell Fluka™
Catalog # HONCR1372-25MG
CAS Number:  
Pesticide reference standard, 2-(Diethylamino)-6-methyl-4-pyrimidinol, neat, Honeywell Fluka™
Catalog # HONCR1372-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • CAS No.:
    42487-72-9
  • Description:
    2-(Diethylamino)-6-methyl-4-pyrimidinol
  • Formula:
    C₉H₁₅N₃O
  • Storage temperature:
    2...8 °C
  • Description:
    2-(Diethylamino)-6-methyl-4-pyrimidinol
  • Freezing point:
    131...133 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results