Order Entry
Germany
ContactUsLinkComponent
Pesticide reference standard, Pentanochlor solution, 100 μg/ml in acetonitrile, Honeywell Fluka™
Pesticide reference standard, Pentanochlor solution, 100 μg/ml in acetonitrile, Honeywell Fluka™
Catalog # HONCR1362-2ML
CAS Number:  
Pesticide reference standard, Pentanochlor solution, 100 μg/ml in acetonitrile, Honeywell Fluka™
Catalog # HONCR1362-2ML
CAS Number:  

Specifications

  • Concentration:
    100 μg/ml in acetonitrile
  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Description:
    Pentanochlor solution
  • Formula:
    C₁₃H₁₈ClNO
  • Storage temperature:
    2...8 °C
  • Description:
    Pentanochlor solution
  • Flash point:
    6 °C
  • Boiling point:
    82 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results