Pharmaceutical analytical standard, β-Estradiol 3-methyl ether solution, 100 μg/ml in acetonitrile, reference material, Honeywell Fluka™
Catalog # HONCR1357-2ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:100 μg/ml in acetonitrile
- Pack type:Glass bottle
- Pk:2 mL
- Description:β-Estradiol 3-methyl ether solution
- Formula:C₁₉H₂₆O₂
- Storage temperature:2...8 °C
- Description:β-Estradiol 3-methyl ether solution
- Flash point:6 °C
- Boiling point:82 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results