Pharmaceutical analytical standard, Cyclopentylalbendazole sulfone, reference material, Honeywell Fluka™
HONCR1319-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1448346-31-3
- Description:Cyclopentylalbendazole sulfone
- Formula:C₁₄H₁₇N₃O₄S
- Storage temperature:Ambient
- Description:Cyclopentylalbendazole sulfone
- Freezing point:273...274 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results