Order Entry
Germany
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Gatifloxacin sesquihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1263-50MG
CAS Number:  
Pharmaceutical analytical standard, Gatifloxacin sesquihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1263-50MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mg
  • CAS No.:
    180200-66-2
  • Description:
    Gatifloxacin sesquihydrate
  • Formula:
    C₁₉H₂₂FN₃O₄ · 1.5H₂O
  • Storage temperature:
    2...8 °C
  • Description:
    Gatifloxacin sesquihydrate
  • Freezing point:
    182 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results