Pharmaceutical analytical standard, Tosufloxacin tosylate, reference material, Honeywell Fluka™
Catalog # HONCR1251-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:115964-29-9
- Description:Tosufloxacin tosylate
- Formula:C₂₆H₂₃F₃N₄O₆S
- Storage temperature:2...8 °C
- Description:Tosufloxacin tosylate
- Freezing point:250...255 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results