Pesticide reference standard, Cythioate, neat, Honeywell Fluka™
Catalog # HONCR1249-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:115-93-5
- Description:Cythioate
- Formula:C₈H₁₂NO₅PS₂
- Storage temperature:2...8 °C
- Description:Cythioate
- Freezing point:71 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results