Pharmaceutical analytical standard, Enoxacin-(piperazinyl-d8) hydrate, reference material, Honeywell Fluka™
Catalog # HONCR1214-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1329642-60-5
- Description:Enoxacin-(piperazinyl-d₈) hydrate
- Formula:C₁₅D₈H₉FN₄O₃ · xH₂O
- Storage temperature:2...8 °C
- Description:Enoxacin-(piperazinyl-d₈) hydrate
- Freezing point:229 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results