Pharmaceutical analytical standard, Oxibendazole-amine hydrochloride, reference material, Honeywell Fluka™
HONCR1213-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1538624-34-8
- Description:Oxibendazole-amine hydrochloride
- Formula:C₁₀H₁₃N₃O · HCl
- Storage temperature:Ambient
- Description:Oxibendazole-amine hydrochloride
- Freezing point:179...181 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results