Order Entry
Germany
ContactUsLinkComponent
 
Pesticide reference standard, Butamifos, neat, Honeywell Fluka™
  HONCR1199-25MG
 :  
Pesticide reference standard, Butamifos, neat, Honeywell Fluka™
  HONCR1199-25MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • CAS No.:
    36335-67-8
  • Description:
    Butamifos
  • Formula:
    C₁₃H₂₁N₂O₄PS
  • Storage temperature:
    2...8 °C
  • Description:
    Butamifos
  • Boiling point:
    422 °C
  • Freezing point:
    25 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results