Order Entry
Germany
ContactUsLinkComponent
Pharmaceutical analytical standard, Benazepril hydrochloride, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Benazepril hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1178-100MG
CAS Number:  
Pharmaceutical analytical standard, Benazepril hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1178-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • CAS No.:
    86541-74-4
  • Description:
    Benazepril hydrochloride
  • Formula:
    C₂₄H₂₈N₂O₅ · HCl
  • Storage temperature:
    Ambient
  • Description:
    Benazepril hydrochloride

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results