Pesticide reference standard, Validamycin A, neat, Honeywell Fluka™
Catalog # HONCR1155-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:37248-47-8
- Description:Validamycin A
- Formula:C₂₀H₃₅NO₁₃
- Storage temperature:2...8 °C
- Description:Validamycin A
- Freezing point:132 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results