Pesticide reference standard, Cloxyfonac, neat, Honeywell Fluka™
Catalog # HONCR1139-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:6386-63-6
- Description:Cloxyfonac
- Formula:C₉H₉ClO₄
- Storage temperature:Ambient
- Description:Cloxyfonac
- Flash point:190.5 °C
- Boiling point:391.4 °C
- Freezing point:140 - 143 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results