Organic analytical standards, Butyl 4-hydroxybenzoate-ring-13C6 solution, 10 μg/mL in acetone, reference material, Honeywell Fluka™
Catalog # HONCR1110-5ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:10 μg/ml in acetone
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:1416711-53-9
- Description:Butyl 4-hydroxybenzoate-ring-¹³C₆ solution
- Formula:¹³C₆C₅H₁₄O₃
- Storage temperature:–20 °C
- Description:Butyl 4-hydroxybenzoate-ring-¹³C₆ solution
- Flash point:–17 °C
- Boiling point:55...57 °C
- Freezing point:–95 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results