Pharmaceutical analytical standard, Cefacetrile sodium, reference material, Honeywell Fluka™
Catalog # HONCR1098-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:23239-41-0
- Description:Cefacetrile sodium
- Formula:C₁₃H₁₂N₃NaO₆S
- Storage temperature:–20 °C
- Description:Cefacetrile sodium
- Freezing point:193 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results