Pesticide reference standard, Dimethachlor Metabolite SYN 528702 sodium salt, neat, Honeywell Fluka™
Catalog # HONCR1093-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:1228182-52-2
- Description:Dimethachlor Metabolite SYN 528702 sodium salt
- Formula:C₁₅H₂₀NNaO₅S
- Storage temperature:2...8 °C
- Description:Dimethachlor Metabolite SYN 528702 sodium salt
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results