Order Entry
Germany
ContactUsLinkComponent
 
Q-VD-OPh
 
Q-VD-OPh

 

  • CAS No.:
    1135695-98-5
  • MDL No.:
    MFCD20527311
  • Protein synonyms:
    Q-Val-Asp-OPh
  • Protein/peptide name:
    Q-VD-OPh
  • Purity:
    95%
  • Molecular weight:
    513,5 g/mol
  • Molecular formula:
    C₂₆H₂₅F₂N₃O₆