Universal vial caps
EKSIA62250
: EKSIGENT
- For:PA 800 plus or PA 800 enhanced instruments
- Description:Universal vial caps
- Pk:100
Universal caps are used in combination with universal vials.
Universal vial caps are designed for use only with CESI 8000, PA 800 Enhanced or PA 800 plus instruments and should not be reused.