Order Entry
Germany
ContactUsLinkComponent
Irritant
3,3'-Diethyloxadicarbocyanine iodide (DODC), high purity
Catalog # 40040.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
3,3'-Diethyloxadicarbocyanine iodide (DODC), high purity
Catalog # 40040.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :486.35 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:14806-50-9
Formula:C₂₃H₂₃IN₂O₂
MDL Number:MFCD00011953