ContactUsLinkComponent
Twin Bucket
Twin Bucket
Catalog # CONT2743S
Supplier:  CONTEC
undefined
Twin Bucket
Catalog # CONT2743S
Supplier:  CONTEC
Specifications
  • Capacity:
    20 L
  • Colour:
    Green
  • Description:
    Twin divided bucket with stainless steel wringer and castors
  • Pk:
    1
Specifications
About this item
Twin Bucket is an autoclavable 20 L divided bucket made of durable PP with a curved stainless steel wringer.
  • Ideal for smaller controlled environments and labs with limited storage space
  • Designed to work with the curved stainless steel wringer
  • Four castors for added mobility
  • Fully autoclavable (except castors)
One side of the bucket can be used for fresh cleaning and disinfecting solutions, and the other one for waste.
Delivery: Supplied with lid, stainless steel wringer and four castors.
Wringer
Product Information