Order Entry
Germany
ContactUsLinkComponent
FIA aromatics standards, ASTM D 1319, Supelco®
FIA aromatics standards, ASTM D 1319, Supelco®
Catalog # SIALCRMFIKR-250ML
Supplier:  Merck
CAS Number:  
FIA aromatics standards, ASTM D 1319, Supelco®
Catalog # SIALCRMFIKR-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    15,18% vol
  • Pk:
    250 mL
  • Description:
    FIA aromatics standard
  • Test method:
    ASTM D 1319

Specifications

About this item

Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for hydrocarbon types in liquid petroleum products by fluorescent indicator adsorption ASTM D1319.