Order Entry
Germany
ContactUsLinkComponent
Iodine 0.5 M I2 (1.0N) volumetric solution, Fluka™
Iodine 0.5 M I2 (1.0N) volumetric solution, Fluka™
Catalog # 319007-500ML
CAS Number:  
Iodine 0.5 M I2 (1.0N) volumetric solution, Fluka™
Catalog # 319007-500ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay:
    0.950 - 1.050 N

Specification Test Results

About this item

 :
 :253.81 g/mol
Density:1.32 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
 :GHS05
 :GHS08
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355