
Ammonium carbonate ≥30.0% (NH₃ basis) ACS, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:506-87-6
:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890
:96.09 g/mol
Merck Index:13,00508
Melting Pt:>59 °C
Density:0.9 g/cm³ (20 °C)
EINECS:208-058-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay of NH₃:Min. 30.0 %
- Non-volatile matter:Max. 0.01 %
- Water insoluble matter:Max. 0.005 %
- Iron (Fe):Max. 5 ppm
- Heavy metals (as Pb):Max. 5 ppm
- Chloride (Cl):Max. 5 ppm
- Total S (as SO₄):Max. 0.002 %