Order Entry
Germany
Orders LinkContactUsLinkComponent
Etching solution according to Murakami

Etching solution according to Murakami

Safety plastic bottle, black, 500 mL
Catalog # KRAF22302.2100
Corrosive
Supplier:   Bernd Kraft
Etching solution according to Murakami
Catalog # KRAF22302.2100
Supplier:  Bernd Kraft
CAS Number:  
Etching solution according to Murakami

Safety plastic bottle, black, 500 mL

CAS Number:  
Price
98,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Suitable for the detection of carbide in annealed chromium steels.

  • UN:1824
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Safety plastic bottle, black
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.