ContactUsLinkComponent
Mini centrifuge / Vortex mixer, FVL-2400N Combi-Spin
Mini centrifuge / Vortex mixer, FVL-2400N Combi-Spin
Catalog # BSAN010202-AAA
Supplier:  BIOSAN
Mini centrifuge / Vortex mixer, FVL-2400N Combi-Spin
Catalog # BSAN010202-AAA
Supplier:  BIOSAN
Specifications
  • Description:
    Mini-centrifuge FVL-2400N Combi–Spin
  • Weight:
    1,7 kg
  • W×D×H:
    190×235×125 mm
  • Pk:
    1
Specifications
About this item
Mini-centrifuge/vortex Combi–Spin FVL-2400N is specially designed for genetic engineering research (for PCR–diagnostics experiments). Units can be used in microbiological, biochemical, clinical laboratories and industrial biotechnological laboratories.
Combi–Spin provides simultaneous mixing and separation of samples, using centrifuge and mixing modules, located on the common spin–module. FVL-2400N is provided with protection mechanism that stops the rotor motion when the lid is opened.