Screw caps for GL 18
Supplier: QVF LABORTECHNIK
Certificates
About this item
Made of the most corrosion resistant, red coloured material PBT.
The cap with the bore are used as connecting cap and the cap without bore are used as sealing cap to close not engaged connections.