Order Entry
Germany
ContactUsLinkComponent
 
3-Iodo-2-propynyl-N-butylcarbamate 97% (by GC)
 :  Merck
 
3-Iodo-2-propynyl-N-butylcarbamate 97% (by GC)
 :  Merck

 

  • Appearance (Colour):
    White to Off-White
  • Appearance (Form):
    Powder or Crystals
  • Infrared Spectrum:
    Conforms to Structure
  • Purity (GC):
    ≥96.5%