
Calcimycin (A23187) free acid ≥98% (by TLC) ionophore
Catalog # 59001.
Supplier: Biotium
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:523.63 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:52665-69-7
Formula:C₂₉H₃₇N₃O₆
MDL Number:MFCD00151202