Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Residual Free Chlorine (RFC), (CRM), Supelco®
Residual Free Chlorine (RFC), (CRM), Supelco®
Residual Free Chlorine (RFC), (CRM), Supelco®
Catalog # QC1450-2ML
Supplier:
Merck
CAS Number:
Certificates
Expand image
Residual Free Chlorine (RFC), (CRM), Supelco®
Catalog # QC1450-2ML
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pk:
2 mL
Description:
Residual Free Chlorine (RFC)
Type:
Certified reference material
Specifications
About this item
Product Details
Documentation
Product Details
Documentation