Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Ammonia - Drinking Water QC, (CRM), Supelco®
Ammonia - Drinking Water QC, (CRM), Supelco®
Catalog # QC1593-2ML
Supplier:
Merck
CAS Number:
Expand image
Ammonia - Drinking Water QC, (CRM), Supelco®
Catalog # QC1593-2ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
2 mL
Description:
Ammonia
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details