Order Entry
Germany
ContactUsLinkComponent
Blood collecting systems, S-Monovette® PFA
Blood collecting systems, S-Monovette® PFA
Catalog # SARS04.1910.001
Supplier:  SARSTEDT
Blood collecting systems, S-Monovette® PFA
Catalog # SARS04.1910.001
Supplier:  SARSTEDT

Specifications

  • Ø:
    13 mm
  • Volume:
    3,8 ml
  • Description:
    S-Monovette® PFA/citrate buffer
  • Length:
    65 mm
  • Seq:
    07
  • Pk:
    50

Specifications

About this item

S-Monovette® is a blood collection system that combines two blood collection techniques – aspiration and vacuum. The S-Monovette® is suitable for all vein conditions and achieves an optimal sample quality, thereby producing the best results. The aspiration technique is a gentle technique for routine blood collection. Using the vacuum technique, a 'fresh' vacuum is always available.

S-Monovette® PFA preparation (3,8% citrate buffer, 0,129 mol/l, pH 5,5) was specially developed for the Siemens Healthineers PFA measuring system, to allow precise analysis of thromocyte function.