Supelco® Organic Reference Standard, Dichloroacetic acid solution
Catalog # CRM47657
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 µg/ml in methyl tert-butyl ether
- Pk:1 mL
- Standards:Certified reference material
- Description:Dichloroacetic acid solution
- Storage conditions:2…8 °C
- Hazard class description:H315
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P280 - P302 + P352 - P332 + P313 - P362 + P364