Supelco® TraceCERT® Organic Reference Standard, Chloroethane solution
Chloroethane solution, 1000 μg/ml in methanol, Certified reference material, TraceCERT®, Ampule of 1 ml, 1 mL
Catalog # CRM40015
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Chloroethane solution
Catalog # CRM40015
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
Caution: P202 - P210 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311
Specifications
Specifications
- Concentration:1000 μg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, TraceCERT®
- Description:Chloroethane solution
- Pk Info:Ampule of 1 ml
- Cat. No.:CRM40015
- M-Clarity quality level:MQ100
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H351 - H370
- MDL No.:MFCD00000961
- Flash point:9,7 °C
- Synonym(s):Chloroethane solution; Ethyl chloride
- Molecular weight:64,51
- CAS No.:75-00-3
- Shipping conditions:WICE
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



