Supelco® Cerilliant® Organic Reference Standard, IGF-1 solution
Catalog # I-033-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml (2% Acetic acid, 2mM Methionine, pH as is), pkg of 1 ml (Cryoules)
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:67763-96-6
- Description:IGF-1 solution
- Storage conditions:–20 °C
- Shipping conditions:DICE
- M-Clarity quality level:MQ300