Coomassie brilliant blue G-250 solution electrophoresis gel stain, ready-to-use
Catalog # 786-497
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:854.04 g/mol
Storage Temperature:Ambient
EINECS:228-058-4
CAS Number:6104-58-1
Formula:C₄₇H₄₈N₃NaO₇S₂
MDL Number:MFCD00078482